missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Applied Biosystems™ ExoSAP-IT™ PCR Product Cleanup Reagent
ExoSAP-IT™ PCR Product Clean-up Reagent is used for enzymatic clean-up of amplified PCR product. It hydrolyses excess primers and nucleotides in a single step.
$224.00 - $7000.00
Specifications
Content And Storage | Shipped on dry ice. Store at -20°C. |
---|---|
Sample Type | PCR Products |
For Use With (Application) | PCR Purification |
Product Line | ExoSAP-IT™ |
Protein Form | Recombinant |
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity | |||||
7820510ML
|
Applied Biosystems™
7820510ML |
5000 Reactions |
Each for $7,000.00
|
|
|||||
782024X1ML
|
Applied Biosystems™
782024X1ML |
2000 Reactions |
Each for $3,105.00
|
|
|||||
782011ML
|
Applied Biosystems™
782011ML |
500 Reactions |
Each for $864.00
|
|
|||||
78200200UL
|
Applied Biosystems™
78200200UL |
100 Reactions |
Each for $224.00
|
|
|||||
Description
- Remove excess primers and unincorporated nucleotides enzymatically
- Prepare DNA sequencing samples with one pipetting step
- Conserve precious PCR products with up to 100% recovery
- Minimize column clean-up and sample loss
Specifications
Shipped on dry ice. Store at -20°C. | |
PCR Purification | |
Recombinant | |
Dry Ice |
PCR Products | |
ExoSAP-IT™ | |
Genetic Diseases |