Gilson™ EXPERT™ Universal Fit Standard Pipette Tips
Feature patented, low-retention polymer that creates a hydrophobic surface to minimize sample binding while dispensing
Supplier: Gilson™ F1733011
Description
- Clear packaging allows easy identification of tips
- Color-coded tip packaging for volume color matching to pipettes
- Graduations tips enable quick-checks when aspirating samples
- Economical and easy-to-use
- Manufactured in sterile, clean-room environment
- Free of Human DNA, RNase, DNase, PCR Inhibitors and Pyrogens
Specifications
Standard Pipette Tips | |
Sterile | |
E200ST Sterilized Tipack | |
Yes | |
Polypropylene |
1 to 200 μL | |
Clear | |
5.144cm | |
EXPERT | |
10, 50, 100μL |