Gilson™ EXPERT™ Universal Fit Standard Pipette Tips
Feature patented, low-retention polymer that creates a hydrophobic surface to minimize sample binding while dispensing
Supplier: Gilson™ F1734511
Description
- Clear packaging allows easy identification of tips
- Color-coded tip packaging for volume color matching to pipettes
- Graduations tips enable quick-checks when aspirating samples
- Economical and easy-to-use
- Manufactured in sterile, clean-room environment
- Free of Human DNA, RNase, DNase, PCR Inhibitors and Pyrogens
Specifications
Standard Pipette Tips | |
Sterile | |
EL10 EasyPack | |
Yes | |
Polypropylene |
0.1 to 10 μL | |
Clear | |
4.534cm | |
EXPERT | |
2, 5, 10μL |