missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ ExpiFectamine™ CHO Transfection Kit

The ExpiFectamine™ CHO Transfection Kit is a core component of the ExpiCHO™ Expression System Kit, designed for transient transfection of high-density cultures of Chinese hamster ovary (CHO) cells.

Supplier:  Gibco™ A29129

 View more versions of this product

Catalog No. A29129


Explore available promotions

Description

Description

The ExpiFectamine CHO Transfection Kit is a core component of the ExpiCHO Expression System Kit (Cat. No. A29133) and enables transient transfection of high-density cultures of Chinese hamster ovary (CHO) cells. The high-efficiency, cationic transfection reagent, coupled with an optimized transfection enhancer and culture feed, will help power the highest possible level of protein expression from ExpiCHO-S Cells (Cat. No. A29127) cultured in ExpiCHO Expression Medium (Cat. No. A2910001).

All components work together synergistically with expression yields of 1–3 g per L of transfected culture demonstrated for some antibody and non-antibody proteins.

Features of the ExpiFectamine CHO Transfection Kit include:

  • Enables robust and reproducible transfection results, giving you greater confidence in your results and ability to use CHO cells from research to discovery
  • Achieve >90% transfection efficiency of high-density suspension CHO cell culture, with matching transfection enhancer and culture feed to boost transfection performance and protein expression
  • Uses one-third to one-half less plasmid DNA than other transfection reagents
  • Scale growth and transfection in culture formats from <1 mL in deep well plates up to 5 L when using Nalgene angled-bottom shake flasks
  • Flexible, reliable culture and transfection protocols with multiple options that readily fit into current transient expression workflows

The ExpiFectamine CHO Transfection Kit is specifically designed to work with ExpiCHO-S Cells in the ExpiCHO Expression System Kit. The transfection kit supplies the cationic ExpiFectamine CHO Transfection Reagent, ExpiFectamine CHO Transfection Enhancer, and ExpiCHO Feed. ExpiCHO-S Cells and ExpiCHO Expression Medium are available separately.

Specifications

Specifications

ExpiCHO-S™, CHO-S, SFM Adapted
Kit includes:
• ExpiFectamine™ CHO Reagent, 3.2 mL
• ExpiFectamine™ CHO Enhancer, 6 mL
• ExpiCHO Feed, 330 mL

Sufficient for transfection of 1 L of culture
Store in the dark at 2°C to 8°C.
Lipid-based Transfection
CHO Transfection Kit
Room Temperature
Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Ovarian Cells
ExpiFectamine™
For 1 L of culture
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.