missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ External-Controlled Hot Plate, 150°C to 371°C, Porcelain-coated Stainless Steel
Enable uniform heating of large volumes and multiple vessels.
Supplier: Thermo Scientific™ RC2240Q
Description
Enable uniform heating of large volumes and multiple vessels using Thermo Scientific™ External-Controlled Hotplates. These hotplates are ideal for a range of applications with acidic or basic aqueous solutions. An external controller operates outside fume hoods and corrosive environment.
Key Features:- Isolation of controls allows operation in hazardous environments
- Large 12 x 12″ (30.5 x 30.5 cm) and 12 x 24″ (30.5 x 60.9 cm) top plate surfaces accommodate up to 40 lb. (18.1 kg) loads
- Power light indicates when control is operating
- Large capacity with excellent temperature stabilityy
- Temperature stability is ±5&°C;C (9&°C;F) over the operating temperature range Embedded heating elements maximize temperature uniformity and evenly transfer heat within ±10&°C;C (18&°C;F) at 150&°C;C (300&°C;F)
- Percentage input controller delivers infinite temperature selection
Construction:
- Porcelain-coated, stainless-steel top offers chemical and stain resistance
- Chemical-resistant cover protects line cord
- Stainless-steel case offers maximum resistance to corrosion
Includes:
5′ (1.5 m) 3-wire cord and plug
Specifications
Hot Plate | |
18 kg | |
Rectangle | |
12 x 24 in. | |
3200 w | |
240 V, 50/60 Hz |
12 in. x 24 in. | |
240 V | |
Porcelain-coated Stainless Steel | |
150°C to 371°C | |
13.3 A | |
50/60 Hz |