Learn More
Mettler Toledo™ F1 OIML Class Standard Weight Set (F1 PL C E)
F1 OIML quality weight set housed in a plastic case includes accessories for handling and cleaning, and a calibration certificate.
Supplier:  Mettler Toledo™ 30406408
Numéro de catalogue. 01912625
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • With calibration certificate
  • Traceability Certificate: Delivered with a Calibration Certificate in accordance with ISO/IEC 17025 requirements
  • Weight Calibration Performed by Robots: State-of-the-art robot technology minimizes errors and increases accuracy
  • Each knob weight has an adjusting cavity, which simplifies weight adjustment without the need for specialized equipment
Spécifications
Spécifications
Test Weight
Plastic
7950 (± 140) kg/m3
Stainless Steel
1 to 500 mg
12 Pieces; With certificate
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS