missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F2R Mouse anti-Human, Clone:5E2E6, Abnova™
Mouse monoclonal antibody raised against recombinant human F2R.
Supplier: Abnova Corporation MAB17583
Description
Coagulation factor II receptor is a 7-transmembrane receptor involved in the regulation of thrombotic response. Proteolytic cleavage leads to the activation of the receptor. F2R is a G-protein coupled receptor family member. [provided by RefSeq]Specifications
| F2R | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunocytochemistry Flow Cytometry Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. |
|
| F2R | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 5E2E6 | |
| coagulation factor II (thrombin) receptor | |
| In PBS (0.05% sodium azide) | |
| CF2R, HTR, PAR1, TR | |
| Recombinant protein corresponding to amino acid 42-176 of human F2R from E. coli. | |
| 2149 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |