Epredia™ Fa-Tech™ Laser Cassette Printer Accessories
Designed for use with Fa-Tech™ Laser Cassette Printers, these accessories will help support all your printing needs.
Supplier: Epredia™ 9001327
Description
VEGA™ Sorter:
- May be purchased with the VEGA™ printer or as an added accessory to an existing VEGA™ printer.
- Organizes batch printing mode for busy mid to large laboratories.
- Achieve prioritized printing as needed outside of the print queue for urgent patient samples.
- Positioned at the base of the VEGA™ printer, keeps the footprint small saving bench space.
- Permits up to 25 cassettes to be pre-printed and sequentially released into the collector tray.
Collector:
- Collection tray for use with Vega Cassette Sorter to keep cassettes in an organized manner.
- Cassettes collected in a vertical fashion for easy visual recognition.
VEGA™ Duo Unit:
- May be purchased with the VEGA™ printer or as an added accessory to an existing VEGA™ printer.
- Partnered with the Vega printer, permits up to 2 users to print jointly in busy laboratories while in both batch and print-on-demand modes keeping printed cassettes organized by user.
- Up to 18 cassettes can be pre-printed and sequentially released into the collection trays keeping up with workflow demand.
- Saves laboratories the need for additional printers lowing capital expense.
Hopper:
- Holds up to 80 additional cassettes for easy exchange to a different color or style of cassettes when using the Nova Laser Printer.
Carousel:
- Utilize six additional hoppers, holding up to a total of 480 cassettes, to be partnered with the Vega Laser Printer.
- Permits for easy replenishment or exchange of cassettes for continual workflow without interruptions for busy laboratories.
Side Delivery System:
- Available for both VEGA™ and NOVA printers.
- Keeps printed cassettes in an organized fashion when printing in either batch or print-on-demand modes.
- Delivers cassettes directly onto collection trays to eliminate workflow delays.
Trays:
- Metal – two trays each holding up to 10 cassettes paired with the Vega Duo Unit to keep cassettes in an organized manner.
- Wooden trays more environmentally friendly.
- Two trays each holding up to 10 cassettes.
Specifications
Hopper Carousel | |
Laser Cassette Printer Accessories | |
Fa-Tech™ VEGA™ Laser Printer |