Epredia™ Fa-Tech™ Laser Cassette Printer Accessories
Designed for use with Fa-Tech™ Laser Cassette Printers, these accessories will help support all your printing needs.
Supplier:  Epredia™ 9001387
 View more versions of this product
Catalog No. 90-013-87
Explore available promotions
Description
Description
VEGA™ Sorter:
  • May be purchased with the VEGA™ printer or as an added accessory to an existing VEGA™ printer.
  • Organizes batch printing mode for busy mid to large laboratories.
  • Achieve prioritized printing as needed outside of the print queue for urgent patient samples.
  • Positioned at the base of the VEGA™ printer, keeps the footprint small saving bench space.
  • Permits up to 25 cassettes to be pre-printed and sequentially released into the collector tray.
Collector:
  • Collection tray for use with Vega Cassette Sorter to keep cassettes in an organized manner.
  • Cassettes collected in a vertical fashion for easy visual recognition.
VEGA™ Duo Unit:
  • May be purchased with the VEGA™ printer or as an added accessory to an existing VEGA™ printer.
  • Partnered with the Vega printer, permits up to 2 users to print jointly in busy laboratories while in both batch and print-on-demand modes keeping printed cassettes organized by user.
  • Up to 18 cassettes can be pre-printed and sequentially released into the collection trays keeping up with workflow demand.
  • Saves laboratories the need for additional printers lowing capital expense.
Hopper:
  • Holds up to 80 additional cassettes for easy exchange to a different color or style of cassettes when using the Nova Laser Printer.
Carousel:
  • Utilize six additional hoppers, holding up to a total of 480 cassettes, to be partnered with the Vega Laser Printer.
  • Permits for easy replenishment or exchange of cassettes for continual workflow without interruptions for busy laboratories.
Side Delivery System:
  • Available for both VEGA™ and NOVA printers.
  • Keeps printed cassettes in an organized fashion when printing in either batch or print-on-demand modes.
  • Delivers cassettes directly onto collection trays to eliminate workflow delays.
Trays:
  • Metal – two trays each holding up to 10 cassettes paired with the Vega Duo Unit to keep cassettes in an organized manner.
  • Wooden trays more environmentally friendly.
  • Two trays each holding up to 10 cassettes.
Specifications
Specifications
Sorter Collector Tray
Laser Cassette Printer Accessories
Fa-Tech™ VEGA™ Laser Printer Sorter