Epredia™ Series 110 Laser Slotted with Front Tab Cassettes with Lids
Manufactured specifically for laser printing, the Epredia™ Series 110 Slotted Cassettes come with separate lids with a tab in the front for easy opening and closing.
$682.11 - $682.11
Specifications
Closure Included | Yes |
---|---|
Quantity | 40 Cassettes/Taped Stack, 25 Stacks/Cs. |
Pore Style | Slots |
Product Type | Cassette |
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
1100110LM
|
Epredia™
1100110LM |
White |
Case of 1000 Each for $682.11
|
|
|||||
1100210LM
|
Epredia™
1100210LM |
Yellow |
Case of 1000 Each for $682.11
|
|
|||||
1100310LM
|
Epredia™
1100310LM |
Blue |
Case of 1000 Each for $682.11
|
|
|||||
1100410LM
|
Epredia™
1100410LM |
Pink |
Case of 1000 Each for $682.11
|
|
|||||
1100510LM
|
Epredia™
1100510LM |
Green |
Case of 1000 Each for $682.11
|
|
|||||
1100610LM
|
Epredia™
1100610LM |
Orange |
Case of 1000 Each for $682.11
|
|
|||||
1100710LM
|
Epredia™
1100710LM |
Gray |
Case of 1000 Each for $682.11
|
|
|||||
1100810LM
|
Epredia™
1100810LM |
Aqua |
Case of 1000 Each for $682.11
|
|
|||||
1100910LM
|
Epredia™
1100910LM |
Lilac |
Case of 1000 Each for $682.11
|
|
|||||
1101210LM
|
Epredia™
1101210LM |
Red |
Case of 1000 Each for $682.11
|
|
|||||
Description
- Epredia™ Laser Cassettes designed for use on both the NOVA™ and VEGA™ Laser Printers
- Contain a special constituent chemical compound that carbonizes when exposed to the laser beam
- Each style comes in different colors with color-matched lids
- Each cassette style is designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing
- Comes in taped stacks of 40 cassettes; 25 stacks per case (for a total of 1000 cassettes with separate lids)
- The Epredia™ Laser Cassette is not compatible with thermal tape style printers
Specifications
Yes | |
40 Cassettes/Taped Stack, 25 Stacks/Cs. | |
Slots | |
Cassette |