Falcon™ Cell Scrapers
Designed to provide maximum accessibility to growth surfaces. Falcon™ Cell Scrapers provide superior scraping ability with dishes, plates, and flasks.
Supplier:  Falcon™ 353085
 View more versions of this product
Catalog No. 08-771-1A
Explore available promotions
Description
Description
  • The improved Falcon Cell Scrapers have been thoughtfully designed to provide maximum accessibility to the growth surfaces in a variety of culture vessels
  • Cross-ribbed polystyrene handle provides greater rigidity to ensure better control while scraping cells
  • Flexibility of the joint between the blade and handle improves ease of access into the neck of a flask or roller bottle Highly compliant thermoplastic elastomer (TPE) blade pivots to provide multiple angles to remove cells from the entire growth surface
  • Available in four sizes—choose the blade and handle length to best accommodate your needs
  • Supplied individually in peel-open, medical-style packaging for sterile presentation
Specifications
Specifications
0.7 in.
For a variety of cell culture vessels, 75 cm&Sup2; Flask
18 cm
100/Cs.
Compliant TPE blade and rigid, cross-ribbed polystyrene handle
1.8 cm
7 in.
1/Pk.
Cell Scraper
Sterile