Falcon™ Sterile Sample Containers
Made of clear polypropylene for strength and visibility. Falcon™ Sterile Sample Containers are ideal for specimen collection.
Supplier:  Falcon™ 354014
 View more versions of this product
Catalog No. 11-838-1
Explore available promotions
Description
Description
  • Sterile, disposable polypropylene containers provide secure sample containment
  • Available in 4 1/2 oz. (110 mL) and 8 oz. (220 mL) sizes both with and without matching lids
  • Feature molded-in graduations for easy measurements
  • Graduated in ounces and milliliters to provide convenient, single-use, timesaving containers for collection, transportation, and storage of a wide variety of specimens
  • Inert and chemically resistant to commonly used laboratory reagents at room temperature
  • Features
    Sterile
  • 4 1/4 oz. graduations from 1/2 oz. to 4 1/2 oz. in 1/4 oz. increments and from 20 mL to 110 mL in 10 mL increments
  • 8 oz. size graduated from 1/4 oz. to 8 oz. in 1/4 oz. increments and from 20 mL to 220 mL in 10 mL increments
Specifications
Specifications
No
Polypropylene
110 mL
20/Pk.
Yes
Sterile
500/Cs.
Videos
SDS
Documents
Documents
Accessories