Learn More
Thermo Scientific Chemicals Fast Green FCF
$64.95 - $496.21
Chemical Identifiers
CAS | 2353-45-9 |
---|---|
Molecular Formula | C37H34N2Na2O10S3 |
Molecular Weight (g/mol) | 808.84 |
MDL Number | MFCD00013053 |
InChI Key | XJBPDZVCYTYRIN-UHFFFAOYSA-L |
Synonym | C.I. 42053 |
PubChem CID | 16887 |
IUPAC Name | disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-5-hydroxybenzenesulfonate |
SMILES | [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1652006
|
Thermo Scientific Chemicals
A1652006 |
5 g |
N/A
|
|
|||||
AAA1652014
|
Thermo Scientific Chemicals
A1652014 |
25 g |
N/A
|
|
|||||
AAA1652022
|
Thermo Scientific Chemicals
A1652022 |
100 g |
N/A
|
|
|||||
Description
Stain for showing collagen, muscle, cytoplasm and cells in mammalian tissuesFast Green FCF is used as a quantitative stain for histones and protein stain in electrophoresis. It is also used as a food dye, which is poorly absorbed by intestine. It is used for tinned green peas and other vegetables, jellies, sauces, fish and desserts. It is considered as a replacement of Light Green SF yellowish in Masson's trichrome. Further, it is utilized in tissue stain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsStain for showing collagen, muscle, cytoplasm and cells in mammalian tissuesFast Green FCF is used as a quantitative stain for histones and protein stain in electrophoresis. It is also used as a food dye, which is poorly absorbed by intestine. It is used for tinned green peas and other vegetables, jellies, sauces, fish and desserts. It is considered as a replacement of Light Green SF yellowish in Masson′s trichrome. Further, it is utilized in tissue stain.
Solubility
Soluble in water, glycerin, glycol and ethanol.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
Chemical Identifiers
2353-45-9 | |
808.84 | |
XJBPDZVCYTYRIN-UHFFFAOYSA-L | |
16887 | |
[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O |
C37H34N2Na2O10S3 | |
MFCD00013053 | |
C.I. 42053 | |
disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-5-hydroxybenzenesulfonate |
Specifications
∼290°C (decomposition) | |
C37H34N2Na2O10S3 | |
5718212 | |
C.I. 42053 | |
XJBPDZVCYTYRIN-UHFFFAOYSA-L | |
disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-5-hydroxybenzenesulfonate | |
16887 | |
Hygroscopic | |
Fast Green FCF |
2353-45-9 | |
MFCD00013053 | |
14,3941 | |
Soluble in water,glycerin,glycol and ethanol. | |
[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O | |
808.84 | |
808.86 | |
5 g |
Safety and Handling
GHS H Statement
H341
Suspected of causing genetic defects.
P201-P202-P280-P308+P313-P501c
H341
EINECSNumber : 219-091-5
RTECSNumber : BQ4425000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only