Learn More
MP Biomedicals™ FastPrep™ Soil Extraction Starter Package
The FastPrep™ Soil Extraction Starter Package includes FastPrep-24 5G instrument, BigPrep 2 x 50 mL Adapter and FastDNA 50 mL SPIN DNA Isolation Kit.
Supplier: MP Biomedicals™ 116005600FP
Description
FastPrep Soil Extraction Starter Package includes:
FastPrep-24™ 5G Instrument (116005500)
The FastPrep-24™ 5G Instrument is a high-speed benchtop homogenizer offering the ultimate in speed and performance for the lysis of biological samples. Indeed, simultaneous homogenization of up to 24 samples takes place within 40 seconds. The FastPrep-24™ 5G Instrument uses a unique, optimized motion to disrupt cells through the multidirectional, simultaneous beating of specialized Lysing Matrix beads on the sample material. Developed for difficult and resistant samples, the FastPrep-24™ 5G Instrument lyses thoroughly and quickly any tissues and cells and thus allows easy and reproducible isolation of stable RNA, active proteins and full-length genomic DNA.
Samples and buffers are added to a 2 mL Lysing Matrix tube containing specialized Lysing Matrix beads. The ergonomic design of the instrument ensures an easy loading of the sample tubes that remains securely sealed during the processing. The homogenization speed and duration times are manually programmed using a touch screen visual display. After setting your speed and time with the touch of a button, just push ''run'', and in less than a minute your samples are completely lysed. The touch screen software based interface also includes a wide range of predefined programs designed for optimal setting of routinely run sample types.
- Intuitive Software, User-Friendly Touch Screen
- Highest speed and Most Power
- Shortest Processing
- Widest range of adapters for sample flexibility
- Highest yield of DNA, RNA and Proteins
- Process any tissue or cellular sample type with highest quality results
BigPrep 2 x 50 mL Adapter (116002525)
- Ambient large-scale adapter that holds 2 x 50 mL lysing matrix tubes
- Frozen samples in lysing matrix tubes stored at -20°C or -80°C are ready for immediate processing
- Made with composite material
- Compatability with FastDNA™ 50 mL pin kit for soil and all maxiprep
FastDNA 50 mL SPIN DNA Isolation Kit for Soil (116560600)
- Specific gravity: Garnet flakes, 4.0 to 4.1
- Sample volume: Up to 5 g per prep
- Matrix type: Garnet
- Molecule enriched: DNA
- Tube compatibiltiy: 50 mL
- Hardness (Mohs): 7.5 to 8
Specifications
Nucleic Acid Extraction and Cell Separation Instrument | |
Touch screen | |
USB Port | |
Approx. 200 μL to 50 mL; up to approx. 25 g | |
52 lb. |
Tissue, Cell Culture, Bacteria | |
FastPrep-24 5G Instrument, BigPrep 2 x 50 mL Adapter and FastDNA 50 mL SPIN DNA Isolation Kit for Soil (1 kit) | |
1 to 120 sec. | |
490 mm | |
23.6 kg |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.