BD FecalSwab™ Collection, Transport and Preservation System of Enteric Bacteria
For the collection of rectal swab and fecal specimens and the preservation of enteric pathogenic bacteria during transport
Supplier:  BD 220258
Catalog No. B220258
Explore available promotions
Description
Description
Supplied in a sterile collection kit format comprising a package containing a tube filled with 2mL of BD FecalSwab transport and preservation medium and a regular size nylon flocked specimen collection swab. The flocked swab applicator can be used to collect the clinical rectal specimen or as a trasferring tool for stool specimens.
Specifications
Specifications
Modified Cary-Blair
5°C to 25°C