Thermo Scientific™ Bovine Serum Albumin (BSA)
Stabilize enzymes, increase PCR yields, and prevent adhesion of enzymes with Thermo Scientific™ Bovine Serum Albumin.
Supplier:  Thermo Scientific™ B14
Catalog No. FERB14
Explore available promotions
Description
Description
Bovine Serum Albumin (BSA) is ideal for stabilization of enzymes during storage and for enzymatic reactions where the absence of nucleases is essential. BSA increases PCR yields from low purity templates. It also prevents adhesion of enzymes to the reaction tubes and tip surfaces.
Highlights
  • Non-modified (non-acetylated)
  • V-fraction
Specifications
Specifications
20 mg/mL
5 mg
Traditional Cloning
Molecular Biology
Bovine Serum Albumin
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.