missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ GeneJET Gel Extraction Kit

Thermo Scientific GeneJET Gel Extraction Kit is designed for rapid and efficient purification of DNA fragments from standard or low-melting point agarose gels run in either TAE or TBE buffer.

$119.00 - $530.00

Specifications

Column Type Spin Column
High-throughput Compatibility Not High-throughput Compatible (Manual)
Test Time 15 min.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
FERK0691
View Documents
Thermo Scientific™
K0691
50 Preps
Each for $119.00
 
FERK0692
View Documents
Thermo Scientific™
K0692
250 Preps
Each for $530.00
 
Description

Description

Thermo Scientific GeneJET Gel Extraction Kit is designed for rapid and efficient purification of DNA fragments from standard or low-melting point agarose gels run in either TAE or TBE buffer.

The kit utilizes a proprietary silica-based membrane technology in the form of a convenient spin column. The kit can be used to purify DNA fragments from 25 bp to 20 kb in size with recovery rates up to 95%. Each GeneJET purification column has a binding capacity of up to 25 μg of DNA and can process up to 1 g of agarose gel. The entire procedure takes just 15 minutes, and the isolated DNA is ready to use in all common downstream applications, including ligation, restriction digestion, PCR, sequencing, and labeling.

Highlights
Highly efficient: up to 95% recoveries in the range of 100 bp to 10 kb
Fast: procedure takes only 15 minutes
Convenient: spin columns are capped and assembled with collection tubes

Applications
Fast and efficient extraction of high purity DNA fragments ideal for use in all conventional molecular biology procedures including:
• FastDigest or conventional restriction digestion
• Automated fluorescent or radioactive sequencing
• PCR
In situ hybridization
• Labeling
• Cloning

Specifications

Specifications

Spin Column
Not High-throughput Compatible (Manual)
15 min.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.