missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ GeneJET PCR Purification Kit

Thermo Scientific GeneJET PCR Purification Kit utilizes a proprietary silica-based membrane technology in the form of a convenient spin column, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions.

$122.00 - $532.00

Specifications

Format Spin Column
High-throughput Compatibility Not High-throughput Compatible (Manual)
Run Time 5 min.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
FERK0701
View Documents
Thermo Scientific™
K0701
50 Preps
Each for $122.00
 
FERK0702
View Documents
Thermo Scientific™
K0702
250 Preps
Each for $532.00
 
Description

Description

Thermo Scientific GeneJET PCR Purification Kit utilizes a proprietary silica-based membrane technology in the form of a convenient spin column, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions. It effectively removes primers, dNTPs, unincorporated labeled nucleotides, enzymes, and salts from PCR and other reaction mixtures.

The kit can be used for purification of DNA fragments from 25 bp to 20 kb with recovery rates up to 100%. Each GeneJET purification column has a total binding capacity of up to 25 μg of DNA, and the entire procedure takes 5 minutes. The purified DNA can be used in common downstream applications, such as sequencing, restriction digestion, labeling, ligation, cloning, in vitro transcription, blotting or in situ hybridization.

Highlights
Highly efficient: 90 to 100% recoveries in the range of 100 bp to 10 kb
Fast: procedure takes only 5 minutes
Convenient: spin columns are capped and assembled with collection tubes

Applications
Fast and efficient purification of DNA fragments ideal for use in all conventional molecular biology procedures including:
• FastDigest or conventional restriction digestion
• Automated fluorescent or radioactive sequencing
• PCR
In vitro transcription
In situ hybridization
• Labeling
• Cloning

Specifications

Specifications

Spin Column
Not High-throughput Compatible (Manual)
5 min.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.