Learn More
Thermo Scientific™ GeneJET Plasmid Maxiprep Kit
Thermo Scientific GeneJET Plasmid Maxiprep Kit, sufficient for 25 preps, utilizes silica-based membrane technology in the form of convenient spin column.
$291.61 - $642.00
Specifications
Concentration | 400 ng/μL |
---|---|
Column Type | Spin Column |
Format | Spin Column |
Isolation Technology | Silica Column |
Sample Type | Bacteria |
Description
GeneJET Plasmid Maxiprep Kit utilizes silica-based membrane technology in the form of convenient spin column. Each prep recovers up to 750 μg of high copy plasmid DNA that can be used in a wide variety of molecular biology procedures, such as restriction endonuclease digestion, PCR, cloning, transformation, automated sequencing, in vitro transcription, and transfection of robust cell lines.
Thermo Scientific GeneJET Plasmid Maxiprep Kit, sufficient for 25 preps, utilizes silica-based membrane technology in the form of convenient spin column. Each prep recovers up to 750 μg of high copy plasmid DNA that can be used in a wide variety of molecular biology procedures, such as restriction endonuclease digestion, PCR, cloning, transformation, automated sequencing, in vitro transcription, and transfection of robust cell lines.
Highlights
• Efficient—high yields up to 750 μg of high quality plasmid DNA from 250 mL of bacterial culture
• High concentration of DNA purified— >400 ng/μL
• Convenient—spin column format, centrifuge or vacuum protocols
• Fast—procedure takes 60 minutes, considerably faster than using gravity flow columns
Applications
Isolated plasmid DNA is suitable for all conventional molecular biology procedures, including:
• Transfection of robust cell lines
• FastDigest restriction enzyme digestion
• Automated fluorescent sequencing
• PCR
• Cloning
• Transformation
• In vitro transcription
Specifications
400 ng/μL | |
Spin Column | |
Bacteria | |
Real-Time Quantitative PCR (qPCR), Transfection, Cloning, Sequencing, In Vitro Transcription, Transformation, Nucleic Acid Labeling, PCR | |
> 200 μg (Large-Scale) Plasmid DNA | |
Plasmid MaxiPrep Kit | |
60 min. |
Spin Column | |
Silica Column | |
Maxi | |
Not High-throughput Compatible (Manual) | |
GeneJET | |
Plasmid DNA |
For Research Use Only. Not for use in diagnostic procedures.