missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ GeneJET Plasmid Miniprep Kit

Thermo Scientific GeneJET Plasmid Miniprep Kit utilizes an exclusive silica-based membrane technology in the form of a convenient spin column.

$114.21 - $446.09

Specifications

Column Type Spin Column
Format Spin Column
Isolation Technology Silica Column
Scale Mini
Sample Type Bacteria
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
FERK0502
View Documents
Thermo Scientific™
K0502
50 Preps
Each for $114.21
 
FERK0503
View Documents
Thermo Scientific™
K0503
250 Preps
Each for $446.09
 
Description

Description

GeneJET Plasmid Miniprep Kit utilizes an exclusive silica-based membrane technology in the form of a convenient spin column. The kit recovers up to 20 μg of high copy plasmid DNA per isolation procedure.

Thermo Scientific GeneJET Plasmid Miniprep Kit utilizes an exclusive silica-based membrane technology in the form of a convenient spin column. The kit recovers up to 20 μg of high copy plasmid DNA per isolation procedure.

Highlights
Efficient—high yields of up to 20 μg of high quality plasmid DNA
Fast—procedure takes less than 14 minutes
Convenient—no phenol/chloroform extraction or alcohol precipitation required
Pure—purified DNA is immediately ready to use

Applications
• Fast isolation of high purity plasmid DNA suitable for all conventional molecular biology procedures, including:
• FastDigest or conventional restriction digestion
• Automated fluorescent and radioactive sequencing
• PCR
In vitro transcription
• Transformation

Principle
A bacterial culture is harvested and lysed. The lysate is then cleared by centrifugation and applied on the silica column to selectively bind DNA molecules at a high salt concentration. The adsorbed DNA is washed to remove contaminants, and the pure plasmid DNA is eluted in a small volume of elution buffer or water. The purified DNA is ready for immediate use in all molecular biology procedures, such as fast and conventional digestion with restriction enzymes, PCR, in vitro transcription, transformation, and automated sequencing. Components Resuspension Solution Lysis Solution Neutralization Solution Wash Solution (concentrated) RNase A Solution Elution Buffer Spin Columns Collection Tubes Detailed Protocol

Specifications

Specifications

Spin Column
Silica Column
Bacteria
Compatible (Vacuum manifold)
GeneJET
Plasmid DNA
Spin Column
Mini
PCR, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, In Vitro Transcription
< 100 μg (Small-Scale) Plasmid DNA
Plasmid MiniPrep Kit
<14 min
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.