missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ GeneRuler 1 kb Plus DNA Ladder

Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels.

$210.00 - $794.00

Specifications

Concentration 6X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Bromophenol Blue, Orange G
Product Type DNA Ladder
Ready to Load No
View More Specs

Products 2
Catalog Number Mfr. No. Kit Contents Price Quantity  
Catalog Number Mfr. No. Kit Contents Price Quantity  
FERSM1331
View Documents
Thermo Scientific™
SM1331
6X DNA Loading Dye, 2 x 1mL
Each for $210.00
 
FERSM1332
View Documents
Thermo Scientific™
SM1332
6X DNA Loading Dye, 10 x 1mL
Each for $794.00
 
Description

Description

GeneRuler 1 kb Plus DNA Ladder 75 to 20,000 bp is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels. The ladder is a mixture of chromatography-purified individual DNA fragments.

GeneRuler 1 kb Plus DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase.

The ladder is supplied with 6X DNA Loading Dye for sample DNA.

Highlights

  • Ideal for both DNA sizing and approximate quantification 
  • Sharp bands 
  • Bright reference bands (given in red)
  • Supplied with loading dye for sample DNA 
Specifications

Specifications

6X
Xylene Cyanol FF, Bromophenol Blue, Orange G
No
Agarose Gels
GeneRuler
Agarose
DNA Ladder
75 to 20000 bp
Sustainable packaging
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.