Learn More
Thermo Scientific™ GeneRuler Low Range DNA Ladder
Perform fast sizing and approximate quantification of low molecular weight double-stranded DNA on agarose gels.
$112.00 - $431.00
Specifications
Concentration | 6X |
---|---|
Gel Type | Agarose |
Label or Dye | Xylene Cyanol FF, Bromophenol Blue, Orange G |
Product Type | DNA Ladder |
Ready to Load | No |
Description
GeneRuler Low Range DNA Ladder is recommended for sizing and approximate quantification of a wide range of double-stranded DNA fragments on agarose or polyacrylamide gels. It is ideal for denaturing polyacrylamide gel electrophoresis. The ladder is a mixture of chromatography-purified individual DNA fragments.
GeneRuler Low Range DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase.
Highlights
- Ideal for both DNA sizing and approximate quantificationÂ
- Ideal for non-denaturing polyacrylamide gel electrophoresisÂ
- Sharp bandsÂ
- Bright reference bands (given in red)
- Supplied with loading dye for sample DNAÂ
Specifications
6X | |
Xylene Cyanol FF, Bromophenol Blue, Orange G | |
No | |
Agarose Gels | |
GeneRuler |
Agarose | |
DNA Ladder | |
25 to 700 bp | |
Sustainable packaging |
For Research Use Only. Not for use in diagnostic procedures.