missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ GeneRuler Ultra Low Range DNA Ladder

Perform sizing and approximate quantification of small dsDNA fragments on agarose or polyacrylamide gels. Ideal for siRNA analysis.

$109.00 - $426.00

Specifications

Concentration 6X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Bromophenol Blue, Orange G
Product Type DNA Ladder
Ready to Load No
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
FERSM1211
View Documents Promotion Details
Thermo Scientific™
SM1211
50 μg
Each for $109.00
 
FERSM1212
View Documents Promotion Details
Thermo Scientific™
SM1212
5 x 50 μg
Each for $426.00
 
Description

Description

GeneRuler Ultra Low Range DNA Ladder is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels. It is ideal for denaturing polyacrylamide gel electrophoresis. The ladder is a mixture of chromatography-purified individual DNA fragments.

GeneRuler Ultra Low Range DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase.

Highlights

  • Ideal for both DNA sizing and approximate quantification
  • Ideal for non-denaturing polyacrylamide gel electrophoresis
  • Ideal for analysis of siRNA
  • Sharp bands
  • Bright reference bands (given in red)
  • Supplied with loading dye for sample DNA
Specifications

Specifications

6X
Xylene Cyanol FF, Bromophenol Blue, Orange G
No
Agarose Gels
GeneRuler
Agarose
DNA Ladder
10 to 300 bp
Sustainable packaging
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.