Learn More
Thermo Scientific™ Lambda DNA/HindIII Marker
Perform sizing and approximate quantification of large double-stranded DNA fragments in agarose gels with this conventional marker composed of 8 sharp bands.
$149.30 - $605.45
Specifications
| Gel Type | Agarose |
|---|---|
| Product Type | DNA/Hind III Marker |
| Ready to Load | No |
| Gel Compatibility | Agarose Gels |
Description
Lambda DNA/HindIII Marker is recommended for sizing of linear double-stranded large DNA fragments in agarose gels. Lambda DNA is digested to completion with the appropriate Thermo Scientitfic restriction enzyme(s) and purified and dissolved in storage buffer. DNA fragments contain various sticky ends depending on the restriction enzyme used for preparation of the marker.
The marker can be labeled radioactively with T4 Polynucleotide Kinase. Alternatively, and it can be labeled radioactively or non-radioactively with Klenow Fragment or Klenow Fragment, exo- with the fill-in reaction.
Highlights
- Sizing and approximate quantification of large DNA fragments
- Sharp bands
- Supplied with loading dye for sample DNA
Specifications
| Agarose | |
| DNA/Hind III Marker | |
| No | |
| Agarose Gels |
For Research Use Only. Not for use in diagnostic procedures.