Thermo Scientific™ pBR322 DNA/BsuRI (HaeIII) Marker
Perform sizing and approximate quantification of small double-stranded DNA fragments in agarose and non-denaturing polyacrylamide gels.
Supplier:  Thermo Scientific™ SM0271
Catalog No. FERSM0271
Explore available promotions
Description
Description
pBR322 DNA/BsuRI (HaeIII) Marker is recommended for sizing and approximate quantification of small linear double-stranded DNA fragments in agarose and non-denaturing polyacrylamide gels. pBR322 DNA is digested to completion with the appropriate Thermo Scientific restriction enzyme, then purified and dissolved in storage buffer. The DNA fragments contain blunt or sticky ends depending on the restriction enzyme used for the marker's preparation
The marker can be labeled radioactively with T4 Polynucleotide Kinase.
Highlights
  • Sizing and approximate quantification of small DNA fragments
  • Sharp bands
  • Supplied with loading dye for sample DNA
Specifications
Specifications
Agarose
No
Sustainable packaging
DNA/BSURI (HAEIII) Marker
Agarose Gels
50 μg
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.