Thermo Scientific™ pUC18 DNA
Optimize cloning with this double-stranded closed circular high copy plasmid DNA, 2686 base pairs with a molecular weight of 1.74 x 106 Da isolated from E.coli.
Supplier: Thermo Scientific™ SD0051
Description
pUC18 and pUC19 vectors are small, high copy number, E. coli plasmids, 2686Â bp in length. They are identical except that they contain multiple cloning sites (MCS) arranged in opposite orientations.
Highlights
- Purified by chromatography using proprietary patented technology
- More than 90% in the supercoiled form
- Isolated from E.coli (dam+, dcm+)
- For pUC18 DNA sequence, pUC19 DNA sequence, sequence analysis and map creation, see free online REviewer tool.
Specifications
Restriction Enzymeâ MCS | |
DNA | |
Cloning | |
50 μg |
0.5 μg/μL | |
DNA | |
DNA | |
pUC18 |
For Research Use Only. Not for use in diagnostic procedures.