Thermo Scientific™ Glycogen, RNA grade
Increase the recovery of nucleic acids during preciptation with this inert carrier.
Supplier:  Thermo Scientific™ R0551
Catalog No. FERR0551
Explore available promotions
Description
Description
Glycogen is a highly purified polysaccharide derived from oysters. It is an inert carrier which significantly increases the recovery of nucleic acids by alcohol precipitation. Glycogen is insoluble in ethanol and forms a precipitate that traps target nucleic acids. During centrifugation, a visible pellet is formed which greatly facilitates handling of the precipitated nucleic acids. Glycogen quantitatively precipitates nucleic acids from diluted solutions with a higher efficiency than that of tRNA, linear polyacrylamide, or sonicated DNA (see References 1-4).
We offer two formulations of aqueous glycogen solutions that allow for fast and efficient DNA or RNA recovery:
Glycogen, RNA grade, can be used for both RNA and DNA precipitation (#R0551).
Glycogen, molecular biology grade, recommended for DNA precipitation (#R0561).
Highlights
  • Ideal for high recovery of oligonucleotides (> 8 bases) and low amounts of DNA/RNA (> 20 pg) from diluted solutions
  • Forms clearly visible pellets
  • Does not interfere with enzymatic reactions†
  • Does not interfere with gel electrophoresis of nucleic acids
  • Does not interfere with spectrophotometrical determination of DNA/RNA concentration, (A260 to 280 measurements)
  • Compatible with DNA transformation and in vitro transfection
Specifications
Specifications
Glycogen
2 x 0.1 mL
Liquid
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.