missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ UTP Solution (100 mM)

Thermo Scientific UTP (uridine 5'-triphosphate) is supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH.

Supplier:  Thermo Scientific™ R0471

Catalog No. FERR0471


Explore available promotions

Description

Description

Thermo Scientific UTP (uridine 5'-triphosphate) is supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH. This highly stable nucleotide is more than 99% pure. UTP can be used in many different molecular biology applications.

Features
• Greater than 99% purity confirmed by HPLC
• Functionally tested in in vitro transcription
• Stable for two years at -20°C and after multiple freeze-thaw cycles

Applications
• aRNA synthesis
• siRNA synthesis
• RNA amplification
in vitro transcription

Specifications

Specifications

100 mM
UTP Solution, 100 mM (0.25 mL)

Store at –20°C.
Unconjugated
UTP Solution
in vitro Transcription, RNA Amplification
Dry Ice
0.25 mL
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.