Ferric Alum Indicator, Volhard, Ricca Chemical
Supplier:  Ricca Chemical Company 305016
 View more versions of this product
Catalog No. 3050-16
Explore available promotions
Description
Description
Specifications
Specifications
Ferric Alum Indicator
7783-83-7,7697-37-2,7732-18-5
LCPUDZUWZDSKMX-UHFFFAOYSA-K
azanium;iron(3+);disulfate;dodecahydrate
197097
Amber
Liquid
Volhard
FeH28NO20S2
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
482.176
Natural Poly Bottle
500 mL
Chemical Identifiers
7783-83-7
482.176
197097
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
FeH28NO20S2
LCPUDZUWZDSKMX-UHFFFAOYSA-K
azanium;iron(3+);disulfate;dodecahydrate