missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ferric ammonium citrate, MP Biomedicals™
Supplier: MP Biomedicals Inc 0215804090
Specifications
Ferric ammonium citrate | |
Brown to Rust | |
C6H11FeNO7+3 | |
ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz | |
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3] | |
264.999 | |
265 |
1185-57-5 | |
7.87g/cm3 at 25°C | |
500 g | |
FRHBOQMZUOWXQL-UHFFFAOYSA-N | |
azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+) | |
118984355 | |
Powder |
Chemical Identifiers
1185-57-5 | |
264.999 | |
ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz | |
azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+) |
C6H11FeNO7+3 | |
FRHBOQMZUOWXQL-UHFFFAOYSA-N | |
118984355 | |
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3] |