missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ferric Chloride, 1% (w/v), Ricca Chemical
$222.65 - $270.41
Specifications
Boiling Point | Approximately 100°C |
---|---|
Melting Point | Approximately 0°C |
Color | Dark Yellow-Orange |
CAS | 7647-01-0,10025-77-1,7732-18-5 |
CAS Max % | 0.09%,0.99%,98.91% |
Quantity | 500 mL |
CAS Min % | 0.09%,0.99%,98.91% |
Packaging | 500 mL Amber Glass Bottle |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-213-119
|
Ricca Chemical Company
R3108000500C |
500 mL |
Each for $222.65
|
|
|||||
01-213-130
|
Ricca Chemical Company
R31080001C |
1 L |
Each for $270.41
|
|
|||||
Specifications
Approximately 100°C | |
Dark Yellow-Orange | |
0.09%,0.99%,98.91% | |
0.09%,0.99%,98.91% |
Approximately 0°C | |
7647-01-0,10025-77-1,7732-18-5 | |
500 mL | |
500 mL Amber Glass Bottle |
Safety and Handling
ShelfLife : 12 Months