missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ferric Chloride CS, USP/EP Yellow Primary Solution, Ricca Chemical
1 mL = 45.0 mg FeCl3.6H2O
Supplier: Ricca Chemical Company 311016
Specifications
Ferric Chloride | |
Yellow Primary Solution, Colorimetric Solution | |
Cl3FeH12O6 | |
NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
iron(3+) hexahydrate trichloride | |
1 mL = 45.0mg FeCl3·6H2O | |
Amber Glass Bottle | |
Liquid |
Yellow | |
10025-77-1,7647-01-0,7732-18-5 | |
MFCD00149712 | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] | |
270.29 | |
EP/USP | |
500 mL |
Chemical Identifiers
10025-77-1 | |
270.29 | |
NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
Cl3FeH12O6 | |
MFCD00149712 | |
iron(3+) hexahydrate trichloride |