missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ferric Nitrate Nonahydrate (Crystalline/Certified ACS), Fisher Chemical™

Supplier:  Fisher Chemical I110100

 View more versions of this product

Catalog No. I110-100



Explore available promotions

Description

Description

Specifications

Specifications

Ferric Nitrate Nonahydrate
7782-61-8
Solid
98.0 to 101.0 %
Fe(NO3)3·9H2O
1466
SZQUEWJRBJDHSM-UHFFFAOYSA-N
iron(3+) nonahydrate trinitrate
16211566
98.0 to 101.0%
Pass test
Negligible
1.680g/cm³
47°C
Purple
100 g
FeH18N3O18
MFCD00149708
ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
403.99
404
Certified ACS
Glass Bottle
<100°C

Chemical Identifiers

7782-61-8
403.99
SZQUEWJRBJDHSM-UHFFFAOYSA-N
16211566
O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
FeH18N3O18
MFCD00149708
ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
iron(3+) nonahydrate trinitrate
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Hygroscopic. Use personal protective equipment. Keep away from clothing and other combustible materials. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:1
Instability:2

DOTInformation : DOT Class 5.1, : Oxidizer

SDS Documents

Documents

Promotions

Promotions