missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ferric Subsulfate, Powder, Purified, Spectrum™ Chemical
$308.47 - $18481.82
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18610538
|
Spectrum Chemical Mfg Cor
F1042125GM |
125 g | Amber Glass Bottle |
Each for $308.47
|
|
||||
18610539
|
Spectrum Chemical Mfg Cor
F1042500GM |
500 g | Amber Glass Bottle |
Each for $910.09
|
|
||||
18610540
|
Spectrum Chemical Mfg Cor
F104225KG |
2.5 kg | Poly Pail |
Each for $2,342.62
|
|
||||
18610541
|
Spectrum Chemical Mfg Cor
F104212KG |
12 kg | Fiber Drum |
Each for $8,011.79
|
|
||||
18610542
|
Spectrum Chemical Mfg Cor
F104245KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
1310-45-8 |
Specifications
1310-45-8 | |
125 g | |
Amber Glass Bottle |
1 | |
Purified |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.