missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ferric Sulfate, MP Biomedicals
Supplier: MP Biomedicals Inc 0215804290
Specifications
Ferric Sulfate | |
500 g | |
ferric sulfate, iron iii sulfate, iron persulfate, iron tersulfate, diiron tris sulphate, diiron trisulfate, ferric persulfate, ferric tersulfate, iron sesquisulfate, ferric sesquisulfate | |
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3] | |
399.858 | |
CHEBI:53438 |
10028-22-5 | |
Fe2O12S3 | |
RUTXIHLAWFEWGM-UHFFFAOYSA-H | |
iron(3+);trisulfate | |
24826 | |
399.9 |
Chemical Identifiers
10028-22-5 | |
399.858 | |
ferric sulfate, iron iii sulfate, iron persulfate, iron tersulfate, diiron tris sulphate, diiron trisulfate, ferric persulfate, ferric tersulfate, iron sesquisulfate, ferric sesquisulfate | |
CHEBI:53438 | |
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3] |
Fe2O12S3 | |
RUTXIHLAWFEWGM-UHFFFAOYSA-H | |
24826 | |
iron(3+);trisulfate |
Safety and Handling
Recommended Storage : Room Temperature