missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ferrocene, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 08720218

 View more versions of this product

Catalog No. AA8720218


Explore available promotions

Description

Description

Ferrocene is used as a catalyst for carbon nanotubes and for rocket propellants. It is also used in the fuel for petrol engines, gasoline antiknock, luminous sensitive materials and exhibit anticancer activity. It acts as a photosensitive catalyst and promote the photochemical degadation of plastic films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
Specifications

Specifications

Ferrocene
173°C to 174°C
249°C
C10H10Fe
50 g
Air and moisture sensitive
ferrocene, bis cyclopentadienyl iron
DFRHTHSZMBROSH-UHFFFAOYSA-N
cyclopenta-1,3-diene;iron
25199998
99%
102-54-5
Yellow to Orange
Characteristic
MFCD00001427
UN1325
14,4037
Insoluble in water. Soluble in organic solvents.
[Fe].c1cccc1.c1cccc1
186.04
186.04
Powder

Chemical Identifiers

102-54-5
186.04
DFRHTHSZMBROSH-UHFFFAOYSA-N
25199998
[Fe].c1cccc1.c1cccc1
C10H10Fe
MFCD00001427
ferrocene, bis cyclopentadienyl iron
cyclopenta-1,3-diene;iron
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H302
Flammable solid.
Harmful if swallowed.

P210-P240-P241-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c

H228-H302

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 203-039-3

RTECSNumber : LK0700000

TSCA : Yes

storageNote1 :

  • Packaged under Argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only