missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferroin indicator solution, Honeywell Fluka™
Redox Indicator, Reag. Ph. Eur., E0 in sulfuric acid 1 mol/l = +1,06 volt
$151.13 - $781.43
Chemical Identifiers
| CAS | 14634-91-4 |
|---|---|
| Molecular Formula | C36H24FeN6O4S |
| Molecular Weight (g/mol) | 692.53 |
| MDL Number | MFCD00036428 |
| InChI Key | CIWXFRVOSDNDJZ-UHFFFAOYSA-L |
| Synonym | ferroin, ferroin indicator solution, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, sulfate 1:1, ferroin sulfate, ferroin solution, 1,10-phenanthroline ferrous sulfate, tris 1,10-phenanthroline-n1,n10 iron sulphate, ferrous 1,10-phenanthroline sulfate, ferroin indicator solution, 25 mmol/l, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, sulfate 1:1 |
| PubChem CID | 84567 |
| SMILES | [Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1 |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 14634-91-4 | |
| 692.53 | |
| CIWXFRVOSDNDJZ-UHFFFAOYSA-L | |
| 84567 |
| C36H24FeN6O4S | |
| MFCD00036428 | |
| ferroin, ferroin indicator solution, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, sulfate 1:1, ferroin sulfate, ferroin solution, 1,10-phenanthroline ferrous sulfate, tris 1,10-phenanthroline-n1,n10 iron sulphate, ferrous 1,10-phenanthroline sulfate, ferroin indicator solution, 25 mmol/l, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, sulfate 1:1 | |
| [Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1 |
Specifications
| 14634-91-4 | |
| C36H24FeN6O4S | |
| MFCD00036428 | |
| ferroin, ferroin indicator solution, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, sulfate 1:1, ferroin sulfate, ferroin solution, 1,10-phenanthroline ferrous sulfate, tris 1,10-phenanthroline-n1,n10 iron sulphate, ferrous 1,10-phenanthroline sulfate, ferroin indicator solution, 25 mmol/l, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, sulfate 1:1 | |
| [Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1 | |
| 692.53 | |
| 692.52g/mol |
| 100 mL | |
| [Fe(C12H8N2)3]SO4 | |
| UN3082 | |
| CIWXFRVOSDNDJZ-UHFFFAOYSA-L | |
| iron(2+);1,10-phenanthroline;sulfate | |
| 84567 | |
| Ferroin indicator solution |
Safety and Handling
DOTInformation : 9 / PGIII