missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Fetal Bovine Serum (FBS), Premium
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
$137.00 - $1292.00
Specifications
Content And Storage | Store at ≤-10°C. |
---|---|
Age | Fetal |
Species | Cattle/Bovine |
Country of Origin | Refer to CoA for specific origin |
Form | Frozen |
Description
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
Sera category: Premium
- Use for general cell culture with common cell lines
- Low endotoxin and hemoglobin levels
- Endotoxin level: <10 EU/mL
- Hemoglobin level: <25 mg/dL
- Origin: refer to CoA for specific origin. All products may not be available in all regions due to importation regulations.
- US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US
Gibco serum delivers
- ISO 13485–certified; processed in FDA registered facilities
- Triple-filtered at 0.1 μm
- Gibco bottle easier to use in hood; helps reduce risk of contamination and improve consistency
- Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
Specifications
Store at ≤-10°C. | |
Cattle/Bovine | |
Frozen | |
Sterile-filtered | |
Premium | |
≤10 EU/mL | |
≤25 mg/dL | |
Dry Ice |
Fetal | |
Refer to CoA for specific origin | |
Fetal Bovine Serum | |
Triple-filtered, 0.1 μm | |
Up to 60 quality tests including 9CFR virus testing, endotoxin, and performance | |
Less waste | |
Premium |