missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Fetal Bovine Serum, Premium Plus

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

$143.00 - $1352.00

Specifications

Content And Storage Store at ≤-10°C.
Age Fetal
Species Cattle/Bovine
Country of Origin Refer to CoA for specific origin
Form Frozen
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Format Price Quantity  
Catalog Number Mfr. No. Quantity Format Price Quantity  
A5669401
View Documents
Gibco™
A5669401
50 mL One Shot
Each for $143.00
 
A5669402
View Documents
Gibco™
A5669402
10 x 50 mL One Shot
Pack of 10 Each for $1,352.00
 
A5669701
View Documents
Gibco™
A5669701
500 mL Bottle
Each for $1,146.00
 
Description

Description

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Sera category: Premium Plus

  • Use for a broad range of cell types, especially sensitive cell lines
  • Our most characterized serum with the lowest endotoxin and hemoglobin levels
  • Endotoxin level: ≤5 EU/mL
  • Hemoglobin level: ≤20 mg/dL
  • Origin: Refer to CoA for specific origin (US, Australia, New Zealand)
  • US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US

Gibco Serum Delivers

  • ISO 13485-certified; processed in FDA-registered facilities
  • Triple filtered at 0.1 μm
  • Gibco bottle easier to use in hood, reduces risk of contamination and helps improve consistency
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
Specifications

Specifications

Store at ≤-10°C.
Cattle/Bovine
Frozen
Standard (Sterile-filtered)
Triple-filtered, 0.1 μm
Triple-filtered, 0.1μm
≤5 EU/mL
≤20 mg/dL
USDA Safety Testing Performed for Australian origins
Fetal
Refer to CoA for specific origin
Fetal Bovine Serum
Sterile
Premium Plus
Up to 70 quality tests including EMA virus testing; USP/EP mycoplasma, endotoxin, performance; biochemical/hormonal profiling; Oritain fingerprinting
Less waste
Dry Ice
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.