missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Fetal Bovine Serum, qualified, Australia DFS Item

Important Update: We've recently introduced Premium Plus Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Certified US origin, New Zealand origin, and Qualified Australia origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.

$2560.95 - $2560.95

Specifications

Age Fetal
Country of Origin Australia
Serum Treatment Standard (Sterile-filtered)
Sterility Sterile
Endotoxin Concentration ≤5 EU/mL
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Format Price Quantity  
Catalog Number Mfr. No. Quantity Format Price Quantity  
10099158 DFS Item
View Documents
Gibco™
10099158
1000 mL Bottle
Each for $2,560.95
 
Description

Description

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera category: Premium (Secure)
  • Origin: Australia.
  • Endotoxin level: ≤5 EU/mL
  • Hemoglobin level: ≤30 mg/dL (levels routinely ≤25 mg/dL)

Aliquot-free 50 mL bottle

The Gibco One Shot 50 mL bottle is designed to eliminate the need to aliquot, and the risk of contamination and variability that comes with it, improving your chances for consistent results.

Features include:

  • Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
  • Engineered to freeze/thaw without splitting or breaking
  • Fits in test tube racks and frees up freezer space
  • No labeling mishaps with our easy-to-read package label containing all of the information you need
  • 33% less waste generated than aliquoting from 500 mL bottles

All products may not be available in all regions due to importation regulations. Contact your local sales representative regarding product availability in your country.

Order Info

Shipping Condition: Dry Ice

Specifications

Specifications

Fetal
Standard (Sterile-filtered)
≤5 EU/mL
Frozen
Australia
Sterile
≤30 mg/dL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'