missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Fetal Bovine Serum, qualified, Canada DFS Item

Introducing the new, aliquot-free Gibco™ One Shot™ FBS 50 mL bottle. Designed to eliminate the need to aliquot, and the risk of contamination and variability that comes with it. And that improves your chances for consistent results.

Supplier:  Gibco™ A3160701

 View more versions of this product

Catalog No. A3160701


Explore available promotions

Description

Description

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera category:Value (Standard)
  • Origin: Canada
  • Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL

Aliquot-free 50 mL bottle

The Gibco One Shot 50 mL bottle is designed to eliminate the need to aliquot, and the risk of contamination and variability that comes with it, improving your chances for consistent results.

Features include:

  • Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
  • Engineered to freeze/thaw without splitting or breaking
  • Fits in test tube racks and frees up freezer space
  • No labeling mishaps with our easy-to-read package label containing all of the information you need
  • 33% less waste generated than aliquoting from 500 mL bottles

All products may not be available in all regions due to importation regulations. Contact your local sales representative regarding product availability in your country.

Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Bovine
One Shot
50 mL
Sterile
Qualified
Frozen
Fetal
Canada
Fetal Bovine Serum
Standard (Sterile-filtered)
Fewer resources used and less waste
One Shot™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions