missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fexofenadine hydrochloride, Thermo Scientific Chemicals

H-1 receptor antagonist

Supplier:  Thermo Scientific Chemicals J63262MB

 View more versions of this product

Catalog No. AAJ63262MB


Explore available promotions

Description

Description

Fexofenadine hydrochloride is used as an antihistamine that inhibits Cox-1, Cox-2, and is a histamine H1 receptor agonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fexofenadine hydrochloride is used as an antihistamine that inhibits Cox-1, Cox-2, and is a histamine H1 receptor agonist.

Solubility
Soluble in Dimethyl sulfoxide (50 mM), and methanol.

Notes
Protect from heat. Store away from oxidizing agents.
Specifications

Specifications

Fexofenadine hydrochloride
148°C to 150°C
C32H40ClNO4
25 mg
fexofenadine hydrochloride, fexofenadine hcl, allegra, telfast, 2-4-1-hydroxy-4-4-hydroxydiphenylmethyl piperidin-1-yl butyl phenyl-2-methylpropanoic acid hydrochloride, fexofenidine hydrochloride, terfenadine carboxylate hydrochloride, mdl 16455 hydrochloride, terfenidine carboxylate hydrochloride
RRJFVPUCXDGFJB-UHFFFAOYNA-N
hydrogen 2-(4-{1-hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butyl}phenyl)-2-methylpropanoic acid chloride
63002
Crystalline Powder
153439-40-8
White
MFCD00865710
14,4068
Soluble in Dimethyl sulfoxide (50 mM),and methanol.
[H+].[Cl-].CC(C)(C(O)=O)C1=CC=C(C=C1)C(O)CCCN1CCC(CC1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
538.13
538.12

Chemical Identifiers

153439-40-8
538.13
RRJFVPUCXDGFJB-UHFFFAOYNA-N
63002
[H+].[Cl-].CC(C)(C(O)=O)C1=CC=C(C=C1)C(O)CCCN1CCC(CC1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
C32H40ClNO4
MFCD00865710
fexofenadine hydrochloride, fexofenadine hcl, allegra, telfast, 2-4-1-hydroxy-4-4-hydroxydiphenylmethyl piperidin-1-yl butyl phenyl-2-methylpropanoic acid hydrochloride, fexofenidine hydrochloride, terfenadine carboxylate hydrochloride, mdl 16455 hydrochloride, terfenidine carboxylate hydrochloride
hydrogen 2-(4-{1-hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butyl}phenyl)-2-methylpropanoic acid chloride
Safety and Handling

Safety and Handling

RTECSNumber : CY1633377

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only