missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fexofenadine hydrochloride, Thermo Scientific Chemicals
H-1 receptor antagonist
Supplier: Thermo Scientific Chemicals J63262MC
Description
Fexofenadine hydrochloride is used as an antihistamine that inhibits Cox-1, Cox-2, and is a histamine H1 receptor agonist.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsFexofenadine hydrochloride is used as an antihistamine that inhibits Cox-1, Cox-2, and is a histamine H1 receptor agonist.
Solubility
Soluble in Dimethyl sulfoxide (50 mM), and methanol.
Notes
Protect from heat. Store away from oxidizing agents.
Specifications
Fexofenadine hydrochloride | |
148°C to 150°C | |
C32H40ClNO4 | |
100 mg | |
fexofenadine hydrochloride, fexofenadine hcl, allegra, telfast, 2-4-1-hydroxy-4-4-hydroxydiphenylmethyl piperidin-1-yl butyl phenyl-2-methylpropanoic acid hydrochloride, fexofenidine hydrochloride, terfenadine carboxylate hydrochloride, mdl 16455 hydrochloride, terfenidine carboxylate hydrochloride | |
RRJFVPUCXDGFJB-UHFFFAOYNA-N | |
hydrogen 2-(4-{1-hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butyl}phenyl)-2-methylpropanoic acid chloride | |
63002 | |
Crystalline Powder |
153439-40-8 | |
White | |
MFCD00865710 | |
14,4068 | |
Soluble in Dimethyl sulfoxide (50 mM),and methanol. | |
[H+].[Cl-].CC(C)(C(O)=O)C1=CC=C(C=C1)C(O)CCCN1CCC(CC1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1 | |
538.13 | |
538.12 |
Chemical Identifiers
153439-40-8 | |
538.13 | |
RRJFVPUCXDGFJB-UHFFFAOYNA-N | |
63002 | |
[H+].[Cl-].CC(C)(C(O)=O)C1=CC=C(C=C1)C(O)CCCN1CCC(CC1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1 |
C32H40ClNO4 | |
MFCD00865710 | |
fexofenadine hydrochloride, fexofenadine hcl, allegra, telfast, 2-4-1-hydroxy-4-4-hydroxydiphenylmethyl piperidin-1-yl butyl phenyl-2-methylpropanoic acid hydrochloride, fexofenidine hydrochloride, terfenadine carboxylate hydrochloride, mdl 16455 hydrochloride, terfenidine carboxylate hydrochloride | |
hydrogen 2-(4-{1-hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butyl}phenyl)-2-methylpropanoic acid chloride |
Safety and Handling
RTECSNumber : CY1633377
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only