Thermo Scientific™ Fiberlite F12-6 x 500 LEX Fixed Angle Rotor with Auto-Lock for LYNX 4000 and 6000 Superspeed Centrifuges
Shorten run set-up time while providing peace of mind that the rotor is secure with this advanced centrifuge rotor.
Supplier:  Thermo Scientific™ 096062375
Catalog No. 096-062375
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
Shorten run set-up time while providing peace-of-mind that the rotor is secure with the Thermo Scientific™ Fiberlite™ F12-6 x 500 Fixed-Angle Rotor, which runs popular 500mL bottles to over 24,400 x g, and supports smaller bottles with available adapters.
Thermo Scientific™ Auto-Lock Rotor Exchange
  • Secure, push-button rotor exchange in less than 3 seconds
  • Improved safety and convenience
  • No tools required
  • Quickly change rotors and applications
Thermo Scientific™ Auto-ID™ Instant Rotor Identification
  • Immediate identification of a rotor
  • Rotor specifications automatically loaded into centrifuge parameters
  • Shortens run set-up time, improves safety
Speed Handle
  • Accelerates and simplifies rotor lid tightening, ensuring lid is properly attached
  • Easier and safer lifting and carrying of rotors, further enhanced with the lightweight design
Specifications
Specifications
Fiberlite F12-6x500 LEX Fixed Angle Rotor
Fixed Angle Rotor
24,471 x g
152 mm
2,302
70 mm
Sorvall LYNX 6000 and 4000 Superspeed Centrifuges
Yes
Carbon Fiber Composite
6 x 500 mL
12,000 rpm
41 mm
160 mm
20.0 lb. (9.1 kg)
20°