3M™ Fibre Gasket, L-149-10
Is part of the welding lens stack. It is placed between the cover plate and the glass shade lens to prevent glass breakage due to heat buildup.
Supplier:  3M™ 70070843365
Catalog No. 29-636-6699
Explore available promotions
Description
Description
  • The gasket is not used with the electronic lens
  • A recommended 3M™ Accessory for select items
Specifications
Specifications
Gray
Gasket
13.97 cm
11.43 x 13.97 cm
10/Pk.
5 in.