missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fibronectin Bovine Protein, Plasma

Catalog No. 33010018
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33010018 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 33010018 Supplier Gibco™ Supplier No. 33010018
Only null left

Fibronectin Bovine Protein, Plasma

Fibronectin is a large, cell surface and plasma protein that exhibits structural and adhesive properties in cell-associated fibrillar matrices. Fibronectin is one of the primary cell adhesion molecules. Bovine Plasma Fibronectin is purified from plasma by affinity chromatography on gelatin-Sepharose and further purified by ion exchange chromatography to remove glycosaminoglycans. Bovine Fibronectin is filtered (0,2μm) and supplied lyophilized.

Specifications

For Use With (Application) Cell Culture
Quantity 1 mg
Gene Alias Fibronectin, Bovine Plasma
Species Cattle/Bovine
Content And Storage Store in freezer (-5 to -30°C).
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Form Solid
Protein Subtype Fibronectin
Research Category Extracellular Matrix & Cell Adhesion, Stem Cell Research

For Research Use Only. Not for use in diagnostic procedures.