missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva Ficoll-Paque™ PLUS Media

Recognized standard in laboratories worldwide for isolation of human lymphocytes from peripheral blood. Cytiva™ Ficoll-Paque™ PLUS Media maintains the viability and a representative distribution of B and T lymphocytes.

$1092.47 - $1092.47

Specifications

Content And Storage 4–30°C
For Use With (Application) Cell Isolation
Endotoxin Level < 0.12 EU/mL
Color Colorless to Slightly Yellow
Product Type Centrifugation Media
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
45001750
View Documents
Cytiva
17144003
6 x 500 mL
Pack of 6 Each for $1,092.47
 
Description

Description

  • Ready-to-use sterile medium
  • Density media optimized for the isolation of lymphocytes from peripheral blood using simple centrifuge-based protocol
  • High yield
  • Maintains viability and representative distribution of B and T lymphocytes
  • Low levels of endotoxin (<0.12 EU/mL)

Specifications

Specifications

4–30°C
< 0.12 EU/mL
Centrifugation Media
Sterile
Cell Isolation
Colorless to Slightly Yellow
1.078 g/mL
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'