Epredia™ Fine Pore Cassettes
For very small biopsy tissue samples, cassettes have a front-edge lid hinge with attached lids that are available in 12 different color varieties
$1883.69 - $1883.69
Specifications
Closure Included | Yes |
---|---|
Packaging Type | Loose Packed |
Quantity | 1000/Cs. |
Pore Style | Fine Pore |
PrintMate Compatible | No |
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
B851735BE
|
Epredia™
B851735BE |
Beige |
Case of 1000 Each for $1,883.69
|
|
|||||
B851735BL
|
Epredia™
B851735BL |
Blue |
Case of 1000 Each for $1,883.69
|
|
|||||
B851735GN
|
Epredia™
B851735GN |
Green |
Case of 1000 Each for $1,883.69
|
|
|||||
B851735GY
|
Epredia™
B851735GY |
Gray |
Case of 1000 Each for $1,883.69
|
|
|||||
B851735LA
|
Epredia™
B851735LA |
Lavender |
Case of 1000 Each for $1,883.69
|
|
|||||
B851735LG
|
Epredia™
B851735LG |
Light Green |
Case of 1000 Each for $1,883.69
|
|
|||||
B851735LI
|
Epredia™
B851735LI |
Lilac |
Case of 1000 Each for $1,883.69
|
|
|||||
B851735OR
|
Epredia™
B851735OR |
Orange |
Case of 1000 Each for $1,883.69
|
|
|||||
B851735TN
|
Epredia™
B851735TN |
Tan |
Case of 1000 Each for $1,883.69
|
|
|||||
B851735WH
|
Epredia™
B851735WH |
White |
Case of 1000 Each for $1,883.69
|
|
|||||
B851735YW
|
Epredia™
B851735YW |
Yellow |
Case of 1000 Each for $1,883.69
|
|
|||||
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifications
Yes | |
1000/Cs. | |
No |
Loose Packed | |
Fine Pore | |
Cassette |