Excelta™ Fine Tip Probes
For microassembly under magnification
Supplier:  Excelta™ 334C
 View more versions of this product
Catalog No. 17467616
Explore available promotions
Description
Description
  • Fine precision
  • Double-ended, stainless steel construction
  • Flame-hardened tips
  • Overall length 5.5 in. (14cm)
Specifications
Specifications
Double Ended Probe
For micro assembly applications under magnifications
45° angled
13.9 cm
Stainless Steel
Hardened tips
5.5 in.