Learn More
Thermo Scientific™ Finnpipette™ F1 Good Laboratory Pipetting (GLP) Kits
Enjoy the convenience of having a variety of Finnpipette™ F1 single channels packaged together, in a range of volumes, with compatible Finntip pipette tips and a pipette stand to support good laboratory pipetting in your lab
$2147.79 - $2868.36
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | ||||||
05-719-507N
|
Thermo Scientific™
4700850N |
1 to 1000 μL |
Each for $2,164.77
|
|
||||||
05-719-508N
|
Thermo Scientific™
4700860N |
0.2 to 1000 μL |
Pack of 4 Each for $2,868.36
|
|
||||||
05-719-509N
|
Thermo Scientific™
4700865N |
10 to 10,000 μL |
Each for $2,147.79
|
|
||||||
05-719-620N
|
Thermo Scientific™
4701060N |
2 to 1000 μL |
Each for $2,164.77
|
|
||||||
Description
Support good laboratory pipetting in your lab with the Thermo Scientific™ Finnpipette™ F1 GLP Kits, which offer a variety of single channels in a range of volumes that are packaged with compatible Finntips, and a convenient pipette stand. Select the kit that is right for you, and take advantage of the Finnpipette F1 performance and lightweight design to maximize your comfort.
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F1: Everything you want in your everyday, go-to pipette
Features:
- Lightweight handle design
- Low plunger and tip attachment & ejection forces
- Secure volume lock to prevent undesirable volume drift during operation
- Silent, effortless volume adjustment
- Adjustable finger rest - comfort for both right- and left-handed users
- Color coded together with Finntips for easier recognition of compatible tips
Kit 1 includes:
- Three Thermo Scientific Finnpipette F1 Pipettes from 1 to 1000 μL:
- 1-10 μL, micro
- 10-100 μL
- 100-1000 μL
- Grip Strip for each handle to improve grip
- Flex tips: 10 micro: 1 x 96; 200: 1 x 96; 1000: 1 x 96
- F-stand
- Good Laboratory Pipetting Guide
- Reagent reservoir demo pack
Kit 2 includes:
- Four Thermo Scientific Finnpipette F1 Pipettes from 0.2 to 1000 μL:
- 0.2-2 μL
- 2-20 μL univ
- 20-200 μL
- 100-1000 μL
- Grip Strip for each handle to improve grip
- Flex tips: 10 micro: 1 x 96; 200: 2 x 96; 1000: 1 x 96
- F-stand
- Good Laboratory Pipetting Guide
- Reagent reservoir demo pack
Kit 3 includes:
- Three Thermo Scientific Finnpipette F1 Pipettes from 10 to 10,000 μL:
- 10-100 μL
- 100-1000 μL
- 1000-10,000 μL
- Grip Strip for each handle to improve grip
- Flex tips: 200: 1 x 96; 1000: 1 x 96; FT 10 mL: 1 x 24
- F-stand
- Good Laboratory Pipetting Guide
- Reagent reservoir demo pack
Kit 4 includes:
- Three Thermo Scientific Finnpipette F1 Pipettes from 20 to 1000 μL:
- 2-20 μL
- 20-200 μL
- 100-1000 μL
- Grip Strip for each handle to improve grip
- Flex tips: 200μL: 2 x 96; 1000 μL: 1 x 96
- F-stand
- Good Laboratory Pipetting Guide
- Reagent reservoir demo pack
Ordering Information:
The kit contains samples racks of Finntip Flex, the most advanced traditional tip offering very low attachment and ejection forces. The Good Laboratory Pipetting Guide contains helpful tips for pipetting including pipetting techniques, ergonomic considerations and how to select tips for best performance.
Warranty: 5 years with web registration.
"
Specifications
1 | |
GLP Kit |